2-Fluorophenylboronic acid

Synonyms: Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid

445223-10G 445223-1G
220067-986EA 398.77 USD
220067-986 220067-987
2-Fluorophenylboronic acid
2-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
MW: 139.92 g/mol
MDL Number: MFCD00674013
CAS Number: 1993-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR