2-Chlorophenylboronic acid

Warning

Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid

445215-1G 445215-5G
220067-984EA 33.78 USD
220067-984 220067-985
2-Chlorophenylboronic acid
2-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
MDL Number: MFCD00674012
CAS Number: 3900-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR