3,5-Dichlorophenylboronic acid

Warning

445207-25G 445207-5G
220067-981EA 394.62 USD
220067-981 220067-982
3,5-Dichlorophenylboronic acid
3,5-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD00051935
CAS Number: 67492-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR