2-Hydroxyacetophenone ≥98%

Warning

Synonyms: 2-Hydroxy-1-phenylethanone, Benzoylcarbinol, Phenacyl hydroxyide, 2-Hydroxyacetophenone, Phenacyl alcohol, α-Hydroxyacetophenone

445134-5G 445134-25G
220067-973EA 106.18 USD
220067-973 220067-972
2-Hydroxyacetophenone ≥98%
2-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 118 - 120 °C (11 -rr)
Melting Pt: 103 - 105 °C
Density: 1.0963 g/cm³ (99 °C)
MDL Number: MFCD00041829
CAS Number: 582-24-1
EINECS: 209-480-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR