1,3-Benzenedisulfonyl chloride ≥97%

Synonyms: Benzene-1,3-di(sulphonyl chloride), Benzene-1,3-disulphonyl chloride

444642-10G 444642-1G
220067-939EA 539.31 USD
220067-939 220067-940
1,3-Benzenedisulfonyl chloride ≥97%
1,3-Benzenedisulphonyl chloride
Formula: C₆H₄Cl₂O₄S₂
MW: 275.13 g/mol
Boiling Pt: 145 °C (1 -rr)
Melting Pt: 63 °C
MDL Number: MFCD00041879
CAS Number: 585-47-7
EINECS: 209-556-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR