Erbium(III) perchlorate 40% (w/w) in aqueous solution ≥99.9% (trace metals basis)

Danger

444111-2G 444111-10G
220067-885EA 90.77 USD
220067-885 220067-884
Erbium(III) perchlorate 40% (w/w) in aqueous solution ≥99.9% (trace metals basis)
Erbium(III) perchlorate
Formula: Cl₃ErO₁₂
MW: 465.61 g/mol
MDL Number: MFCD00016073
CAS Number: 14017-55-1
EINECS: 237-840-4
UN: 3211
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR