1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥97%

Synonyms: 1,1,1,3,3,3-Hexafluoro-2-methylpropan-2-ol, Hexafluoro-tert-butanol, Hexafluoro-2-methylisopropanol

444014-5G 444014-10G
220067-876EA 77.54 USD
220067-876 220067-875
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥97%
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Formula: C₄H₄F₆O
MW: 182.07 g/mol
Boiling Pt: 60.5…61.5 °C (1013 hPa)
Density: 1.484 g/cm³ (20 °C)
MDL Number: MFCD00039259
CAS Number: 1515-14-6
EINECS: 216-155-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR