4-Bromo-1,1,2-trifluoro-1-butene ≥97%

443743-5ML 443743-25ML
220067-844EA 80.12 USD
220067-844 220067-843
4-Bromo-1,1,2-trifluoro-1-butene ≥97%
4-Bromo-1,1,2-trifluoro-1-butene
Formula: C₄H₄BrF₃
MW: 188.97 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.639 g/cm³ (20 °C)
MDL Number: MFCD00039274
CAS Number: 10493-44-4
EINECS: 234-019-2
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR