2,4,4-Trimethyl-1-pentene ≥96%

Danger

443018-25ML 443018-100ML
220067-835EA 65.69 USD
220067-835 220067-834
2,4,4-Trimethyl-1-pentene ≥96%
2,4,4-Trimethyl-1-pentene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –93.48 °C
Density: 0.71509 g/cm³ (20 °C)
Flash Pt: –5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008855
CAS Number: 107-39-1
EINECS: 203-486-4
UN: 2050
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR