2-Bromobenzenesulfonyl chloride ≥97%

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

442844-1G 442844-5G
220066-520EA 33.38 USD
220066-520 220066-521
2-Bromobenzenesulfonyl chloride ≥97%
2-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52
Boiling Pt: 127…128 °C (3.6 mmHg)
Melting Pt: 51 °C
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR