4-Dodecylbenzenesulfonic acid mixture of isomers ≥95%

Danger

Synonyms: P-Dodecylbenzenesulphonic acid

44198-1L 44198-250ML
220065-991EA 227.23 USD
220065-991 220065-992
4-Dodecylbenzenesulfonic acid mixture of isomers ≥95%
4-Dodecylbenzenesulfonic acid
Formula: C₁₈H₃₀O₃S
MW: 326.49 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 10 °C
Density: solvent 0.9 g/cm³ (25 °C)
MDL Number: MFCD00066677
CAS Number: 121-65-3
EINECS: 204-489-3
UN: 2586
ADR: 8,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR