Barium ≥99.9%

441880-25G 441880-5G
220067-772EA 1707.69 USD
220067-772 220067-773
Barium ≥99.9%
Barium
Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR