Calcium ≥99.99%

Danger

441872-25G 441872-5G
220067-770EA 1861.54 USD
220067-770 220067-771
Calcium ≥99.99%
Calcium
Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 231-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR