3-Formylphenylboronic acid

Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid

441651-25G 441651-5G
220066-391EA 44.17 USD
220066-391 220066-392
3-Formylphenylboronic acid
3-Formylphenylboronic acid
Formula: C₇H₇BO₃
MW: 149.94 g/mol
MDL Number: MFCD00161356
CAS Number: 87199-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR