3-Fluorophenylboronic acid

Warning

Synonyms: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

441643-10G 441643-1G
220066-387EA 205.62 USD
220066-387 220066-388
3-Fluorophenylboronic acid
3-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00236042
CAS Number: 768-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR