Tetrahydrofuran-[D₈]

Danger

441406-10ML 441406-5X.5ML
220067-754EA 176.92 USD
220067-754 220067-757
Tetrahydrofuran-[D₈]
Tetrahydrofuran-[D₈]
Formula: C₄D₈O
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: -108 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: -17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR