2-Bromo-4,5-dimethoxybenzoic acid ≥98%

Warning

Synonyms: 6-Bromoveratric acid

441074-5G 441074-1G
220067-659EA 252 USD
220067-659 220067-658
2-Bromo-4,5-dimethoxybenzoic acid ≥98%
2-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.07 g/mol
MDL Number: MFCD00017542
CAS Number: 6286-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR