Octyltriethoxysilane

Warning

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

440213-100ML 440213-1L
220067-576EA 80.03 USD
220067-576 220067-577
Octyltriethoxysilane
Octyltriethoxysilane
Formula: C₁₄H₃₂O₃Si
MW: 276.49 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.8515 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039883
CAS Number: 2943-75-1
EINECS: 220-941-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR