(3-Glycidoxypropyl)trimethoxysilane

Synonyms: (3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane

440167-1L 440167-100ML 440167-500ML
220067-569EA 447.23 USD
220067-569 220067-568 220067-570
(3-Glycidoxypropyl)trimethoxysilane
(3-Glycidoxypropyl)trimethoxysilane
Formula: C₉H₂₀O₅Si
MW: 236.34 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005144
CAS Number: 2530-83-8
EINECS: 219-784-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR