Trichloro(phenyl)silane

Danger

Synonyms: Phenyltrichlorosilane

440108-1L 440108-100ML
220067-559EA 180 USD
220067-559 220067-558
Trichloro(phenyl)silane
Trichloro(phenyl)silane
Formula: C₆H₅Cl₃Si
MW: 211.55 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.315 g/cm³ (20 °C)
MDL Number: MFCD00000479
CAS Number: 98-13-5
EINECS: 202-640-8
UN: 1804
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR