Gadolinium trichloride, anhydrous ≥99.99%

439770-25G 439770-5G
220067-526EA 1187.69 USD
220067-526 220067-527
Gadolinium trichloride, anhydrous ≥99.99%
Gadolinium trichloride
Formula: Cl₃Gd
MW: 263.61 g/mol
Melting Pt: 604 - 614 °C
Density: 4.52 g/cm³ (0 °C)
MDL Number: MFCD00011024
CAS Number: 10138-52-0
EINECS: 233-386-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR