Strontium chloride, anhydrous ≥99.99%

Warning

439665-25G 439665-5G
220067-513EA 576.92 USD
220067-513 220067-514
Strontium chloride, anhydrous ≥99.99%
Strontium chloride
Formula: Cl₂Sr
MW: 158.53 g/mol
Boiling Pt: 1250 °C (1013 hPa)
Melting Pt: 868 °C
Density: 1.96 g/cm³ (20 °C)
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR