Terbium(III) chloride, anhydrous ≥99.9%

439657-10G 439657-2G
220067-511EA 718.46 USD
220067-511 220067-512
Terbium(III) chloride, anhydrous ≥99.9%
Terbium(III) chloride
Formula: Cl₃Tb
MW: 265.28 g/mol
Melting Pt: 588 °C
Density: 4.35 g/cm³ (25 °C)
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR