Thulium(III) chloride, anhydrous ≥99.9%

439649-1G 439649-5G
220066-816EA 83.77 USD
220066-816 220067-510
Thulium(III) chloride, anhydrous ≥99.9%
Thulium(III) chloride
Formula: Cl₃Tm
MW: 275.29 g/mol
Melting Pt: 820.85 °C
MDL Number: MFCD00011283
CAS Number: 13537-18-3
EINECS: 236-904-9

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR