4-(Trifluoromethyl)phenylboronic acid

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

439320-5G 439320-1G
220067-507EA 114.92 USD
220067-507 220067-506
4-(Trifluoromethyl)phenylboronic acid
4-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR