Potassium iodate ≥99.998%

Warning

438464-25G 438464-5G
220067-427EA 393.85 USD
220067-427 220067-428
Potassium iodate ≥99.998%
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR