Tetrahydrofuran-[D₈]

Danger

437727-1G 437727-10G 437727-5G
220067-352EA 25.66 USD
220067-352 220067-351 220067-353
Tetrahydrofuran-[D₈]
Tetrahydrofuran-[D₈]
Formula: C₄D₈O
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: -108 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: -17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR