Ethylbenzene-[D10]

Synonyms: Phenylethane-D10

437344-10ML 437344-1ML 437344-5ML
220067-311EA 94.46 USD
220067-311 220067-312 220067-313
Ethylbenzene-[D10]
Ethylbenzene-[D10]
Formula: D₃CCD₂OC₆D₅
MW: 116.09 g/mol
Boiling Pt: 134.5 °C (1013 hPa)
Density: 0.94 g/cm³ (20 °C)
CAS Number: 25837-05-2
EINECS: 247-292-8
UN: 1175
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR