2-Thiopheneboronic acid

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

436836-5G 436836-1G
220067-268EA 53.45 USD
220067-268 220067-267
2-Thiopheneboronic acid
2-Thiopheneboronic acid
Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 146 - 147 °C
MDL Number: MFCD00151850
CAS Number: 6165-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR