Lutetium trinitrate hydrate ≥99.9%

Synonyms: Lutetium (III) nitrate hydrate

436429-1G 436429-5G
220067-259EA 122 USD
220067-259 220067-260
Lutetium trinitrate hydrate ≥99.9%
Lutetium trinitrate hydrate
Formula: LuN₃O₉·H₂O
MW: 379 g/mol
MDL Number: MFCD00150421
CAS Number: 100641-16-5
EINECS: 233-241-7
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR