4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98%

436097-1G 436097-5G
220065-010EA 530.77 USD
220065-010 220065-011
4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98%
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00061313
CAS Number: 90176-80-0
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR