Benzamidine hydrochloride ≥99%

Warning

Synonyms: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

434760-5G 434760-25G
220067-102EA 88.48 USD
220067-102 220067-101
Benzamidine hydrochloride ≥99%
Benzamidine hydrochloride
Formula: C₇H₉ClN₂
MW: 156.61 g/mol
Melting Pt: 174…179 °C
MDL Number: MFCD00066285
CAS Number: 1670-14-0
EINECS: 216-795-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR