1-Ethyl-3-methylimidazoliumthiocyanate, Sigma-Aldrich®

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate

43437-1KG-F 43437-100G-F
220067-072EA 2920 USD
220067-072 220067-071
1-Ethyl-3-methylimidazoliumthiocyanate, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumthiocyanate
Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
MDL Number: MFCD06798182
CAS Number: 331717-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR