Magnesium sulfate monohydrate ≥97%

Synonyms: Magnesium sulphate hydrate

434183-500G 434183-1KG
220067-056EA 176.92 USD
220067-056 220067-055
Magnesium sulfate monohydrate ≥97%
Magnesium sulphate monohydrate
Formula: MgSO₄·1H₂O
MW: 138.38 g/mol
Density: 2.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149787
CAS Number: 14168-73-1
EINECS: 231-298-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR