(±)-1-tert-Butoxy-2-propanol ≥99%

Synonyms: 1-Methyl-2-(tert-butoxy)ethanol, (±)-Monopropylene glycol tert-butyl ether, (±)-Propylene glycol mono-tert-butyl ether, Propylene glycol 1-tert-Butyl ether, ARCOSOLV® PTB, 1-(tert-Butoxy)-2-propanol, 1-tert-Butoxy-2-propanol

433845-250ML 433845-1L
220067-015EA 153.69 USD
220067-015 220067-014
(±)-1-tert-Butoxy-2-propanol ≥99%
(±)-1-tert-Butoxy-2-propanol
Formula: C₇H₁₆O₂
MW: 132.2 g/mol
MDL Number: MFCD00077674
CAS Number: 57018-52-7
EINECS: 406-180-0
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR