3-(o-Tolyl)propionic acid ≥99%

Synonyms: 3-(2-Methylphenyl)propionic acid, 2-Methylhydrocinnamic acid, 3-(2-Methylphenyl)propanoic acid

433330-1G 433330-10G
220066-992EA 48.46 USD
220066-992 220066-991
3-(o-Tolyl)propionic acid ≥99%
3-(o-Tolyl)propionic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00079773
CAS Number: 22084-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR