5-Bromo-1-indanone

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one

433098-1G 433098-5G
220066-982EA 26.92 USD
220066-982 220066-983
5-Bromo-1-indanone
5-Bromo-1-indanone
Formula: C₉H₇BrO
MW: 211.06 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 126 - 127 °C
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR