2,4-Dimethoxybenzylamine ≥98%

432725-25ML 432725-5ML
220066-966EA 204.62 USD
220066-966 220066-967
2,4-Dimethoxybenzylamine ≥98%
2,4-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MDL Number: MFCD00052393
CAS Number: 20781-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR