Sodium amide ≥95%

Danger

432504-25G 432504-100G
220066-961EA 34.95 USD
220066-961 220066-960
Sodium amide ≥95%
Sodium amide
Formula: H₂NNa
MW: 39.01 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011117
CAS Number: 7782-92-5
EINECS: 231-971-0
UN: 1390
ADR: 4.3,II
Merck Index: 12,08717

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR