2-Formylphenylboronic acid

Synonyms: 2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane

431958-1G 431958-5G
220066-907EA 31.71 USD
220066-907 220066-908
2-Formylphenylboronic acid
2-Formylphenylboronic acid
Formula: C₇H₇BO₃
MW: 149.94 g/mol
MDL Number: MFCD00151822
CAS Number: 40138-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR