Potassium hydrogen carbonate ≥99.99%

431583-50G 431583-250G
220066-875EA 184.62 USD
220066-875 220066-874
Potassium hydrogen carbonate ≥99.99%
Potassium hydrogen carbonate
Formula: KHCO₃
Melting Pt: 292 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR