Lithium perchlorate ≥99.99%

Danger

431567-50G 431567-250G
220066-871EA 315.38 USD
220066-871 220066-870
Lithium perchlorate ≥99.99%
Lithium perchlorate
Formula: ClLiO₄
MW: 106.39 g/mol
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 236…237 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011079
CAS Number: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1,II
Merck Index: 13,05561

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR