Tin(II) chloride dihydrate ≥99.99%

Danger

Synonyms: Stannous chloride dihydrate

431508-250G 431508-50G
220066-863EA 290.77 USD
220066-863 220066-864
Tin(II) chloride dihydrate ≥99.99%
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR