Disodium hydrogen phosphate heptahydrate ≥99.99%

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

431478-50G 431478-250G
220066-861EA 57.38 USD
220066-861 220066-860
Disodium hydrogen phosphate heptahydrate ≥99.99%
Disodium hydrogen phosphate heptahydrate
Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR