Dodecamolybdophosphoric acid hydrate

Danger

Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

431400-10G 431400-50G
220066-786EA 164.31 USD
220066-786 220066-787
Dodecamolybdophosphoric acid hydrate
Dodecamolybdophosphoric acid hydrate
Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78 - 90 °C
Density: 1.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3260
ADR: 8,III
Merck Index: 12,07498
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f3351423346c386864e8e79e025bbb8578de04&vid=f8f3351423346c386864e8e79e025bbb8578de04&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR