(Benzyl)triphenylphosphonium bromide

Synonyms: Benzyltriphenylphosphonium bromide

430056-50G 430056-250G
220066-727EA 113.08 USD
220066-727 220066-726
(Benzyl)triphenylphosphonium bromide
(Benzyl)triphenylphosphonium bromide
Formula: C₂₅H₂₂BrP
MW: 433.33 g/mol
Melting Pt: 299…301 °C
MDL Number: MFCD00031707
CAS Number: 1449-46-3
EINECS: 215-908-4
UN: 3464
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR