2-Amino-6-fluorobenzonitrile ≥99%

Synonyms: 2-Cyano-3-fluoroaniline

429856-10G 429856-1G
220066-707EA 472.77 USD
220066-707 220066-708
2-Amino-6-fluorobenzonitrile ≥99%
2-Amino-6-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 127 °C
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR