Lithium chloride, anhydrous ≥99.998%

Warning

429457-25G 429457-5G
220066-681EA 407.69 USD
220066-681 220066-682
Lithium chloride, anhydrous ≥99.998%
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR