Zinc chloride, anhydrous

Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

429430-5G 429430-25G
220066-676EA 136.77 USD
220066-676 220066-675
Zinc chloride, anhydrous
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR