α-Bromo-3-iodotoulene ≥95%

Danger

Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide

427691-25G 427691-5G
220066-552EA 401.69 USD
220066-552 220066-553
α-Bromo-3-iodotoulene ≥95%
α-Bromo-3-iodotoulene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00019016
CAS Number: 49617-83-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR